Would you like to log in again?
Shipper, Multisample EAS Ambient, Therapak®
Supplier: Therapak
56883
10029-416CS 104.53 USD InStock
10029-416
Shipper, Multisample EAS Ambient, Therapak®
Mailers and Shippers
Large capacity ambient shipping system accommodates multiple specimen transport bags for consolidation at collection site and shipping to testing laboratory
Features a pop-up style box that is easy to assemble. Easy-fold flaps on both ends of the box enable the shipper to easily slide the included specimen bag and absorbent containing sample tubes inside the box prior to securing the flaps for shipment. Includes an Exempt Animal Specimen label and biohazard symbol label.
System includes a large transport box (ID: 11 x 10.5 x 4"; OD: 11.25 x 10.75 x 4.25") with easy lock flaps on both ends of the box, a 13 x 18" biohazard transport bag, and 12 x 12" absorbent sheet. Holds up to ten 6 x 9" specimen transport bags.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty