Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acridinium C2 NHS Ester

Supplier: Enzo Life Sciences
Synonyms: 4-(2-Succinimidyl-oxycarbonylethyl)-phenyl-10-acridinium-9-carboxylate Triflurormethyl sulphonateAcridinium C2 N-hydroxysuccinimide ester4-(2-Succinimidyl-oxycarbonylethyl)-phenyl-10-acridinium-9-carboxylate Triflurormethyl Sulfonate

ADI-906-000-1
89141-656EA 484 USD InStock
89141-656
Acridinium C2 NHS Ester
Acridinium C2 NHS Ester
Species independent, solid.


Formula: C₂₈H₂₃N₂O₆·CF₃O₃S
MW: 632.56 g/mol
CAS Number: 177332-37-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/15/45347415.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty