Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMTV Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-P13S 100-401-P13S
RL100-401-P13SEA 147.5 USD InStock
CA100-401-P13S RL100-401-P13S
Anti-MMTV Rabbit Polyclonal Antibody
Antibodies
Anti-MMTV Antibody has been tested for use in western blotting. Specific conditions for reactivity should be optimized by the end user. Expect band approximately 41.2 kDa in size corresponding to MMTV surface Protein by western blotting.

Recommended Dilutions: ELISA: 1:10000-1:20000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: MMTV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty