Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PGMB Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-103-237S 200-103-237S
RL200-103-237SEA 148 USD InStock
CA10036-822 RL200-103-237S
Anti-PGMB Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Beta-Phosphoglucomutase has been assayed against 1.0 µg of b-Phosphoglucomutase (Lactococcus lacti) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:1000 - 1:5000;
Do not add Sodium Azide!

Type: Primary
Antigen: PGMB
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Lactococcus lacti

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty