Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGAX Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 2H7]

Supplier: Rockland Immunochemicals
200-308-N63 200-308-N63
RL200-308-N63EA 467 USD InStock
RL200-308-N63 CA10036-940
Anti-ITGAX Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 2H7]
Antibodies
Anti-CD11c is useful for Flow Cytometry, Immunofluorescence, and Immunoprecipitation. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5µl/test/1x10e5 to 1x10e8 cells;
Do not freeze! Light sensitive!

Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: 2H7
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty