Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CPDA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

200-4352S 200-4352S
RL200-4352SEA 148 USD InStock
CA10037-070 RL200-4352S
Anti-CPDA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Carboxypeptidase A is useful in ELISA, western blot, and immunochemistry. Optimal titers are to be reached by researchers.

Recommended Dilutions: ELISA: 1:3000 - 1:10000; Western Blot: 1:300 - 1:2000;
Do not add Sodium Azide!

Type: Primary
Antigen: CPDA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty