Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SELL Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MEL-14]

Supplier: Rockland Immunochemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

200-508-N87 200-508-N87
RL200-508-N87EA 539 USD InStock
RL200-508-N87 CA200-508-N87
Anti-SELL Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MEL-14]
Antibodies
Anti-CD62L is useful for immunochemistry, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: MEL-14
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty