Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STIM2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-J50 600-401-J50
RL600-401-J50EA 518 USD InStock
RL600-401-J50 CA10037-268
Anti-STIM2 Rabbit Polyclonal Antibody
Antibodies
Anti-Stim II antibody is useful for ELISA, immunohistochemistry, and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~84kDa corresponding to the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1µg/ml

Type: Primary
Antigen: STIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty