Would you like to log in again?
PGE1 ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-005
89141-058EA 537.7 USD InStock
89141-058
PGE1 ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of PGE1 in culture supernatants, plasma, serum, saliva, and urine from any species
The PGE1 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405nm. Assay up to 38 samples in duplicate per kit with excellent detection sensitivity.
Cited sample types include simulated interstitial lung fluid.
Ordering information: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-274 (5x96 wells)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty