Would you like to log in again?
Direct cGMP ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-014
89141-068EA 750.48 USD InStock
89141-068
Direct cGMP ELISA Kit, Enzo Life Sciences
Assays ELISAs
Most sensitive (0.025pmol/m) colorimetric kit for intracellular cGMP quantification.
For the quantitative determination of cGMP in cell lysates and tissue from any species. Cited sample types include cerebral spinal fluid and microdialysate.
This is a colorimetric competitive immunoassay kit for the quantitation of intracellular cGMP in cells or tissues lysed in 0.1 M HCl, which is used to stop endogenous phosphodiesterase activity and stabilize the released cyclic GMP. The 0.1M HCl treated samples are then analyzed directly in a microtiter plate without extraction, drying and reconstitution. Sensitivity is increased by acetylation (reagents included). Absorbance is read at 405 nm.The ready-to-use liquid color-coded reagents provided reduce error.
Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, 0.1M HCl, Neutralizing reagent, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Acetic anhydride, Triethylamine. Alternative size available: 89141-284 (5x96 wells).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty