Would you like to log in again?
Urinary Prostacyclin ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-025
89141-084EA 537.7 USD InStock
89141-084
Urinary Prostacyclin ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of urinary prostacyclin metabolites in culture supernatants and urine from any species.
The Urinary Prostacyclin EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 2 by measuring the stable metabolites in one simple assay.
Ordering information: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-292 (5x96 wells)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty