Would you like to log in again?
Protein A ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-057
89141-106EA 911.06 USD InStock
89141-106
Protein A ELISA Kit, Enzo Life Sciences
Assays ELISAs
Most sensitive (9.01pg/ml) colorimetric ELISA for monitoring residual contamination.
For the quantitative determination of residual protein A contamination in protein A purified IgG preparations from any species.
The Protein A EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Ultra-sensitive quantification (< 1 ppm) of Protein A residuals in purified humanized monoclonal antibody preparations. This kit recognizes four different Protein A constructs.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate,Standard, TMB Substrate, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty