Would you like to log in again?
[pThr180/Tyr182]p38 ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-101
89141-154EA 877.04 USD InStock
89141-154
[pThr180/Tyr182]p38 ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of [pThr180/Tyr182]p38 in cell lysates of human and mouse origin.
The [pThr180/Tyr182]p38 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450nm. Use less precious sample per test compared to Western blot with higher throughput sample analysis; measure up to 40 samples in duplicate per kit.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 4 concentrate, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty