Would you like to log in again?
β-Catenin ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-135
89141-192EA 880.08 USD InStock
89141-192
β-Catenin ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of β-Catenin in cell lysates of human, mouse, and rat origin.
The β-Catenin EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours that measures pan β-Catenin. Absorbance is read at 450nm. This kit has been validated in-house with human, mouse, and rat cell lysates and correlated Western blot. The β-Catenin EIA kit detects as little 27pg/ml β-catenin surpassing semi-quantitative Western blot analysis and provides simple liquid color-coded reagents that reduce laboratory error.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, RIPA Cell lysis buffer 2, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty