Would you like to log in again?
IGF-1 (human), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-150
89141-216EA 766.88 USD InStock
89141-216
IGF-1 (human), ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive ELISA, measuring as little as 50pg/ml of IGF-1 in just 4 hours.
For the quantitative determination of human IGF-1 in plasma and serum.
The IGF-1 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4 hours. Absorbance is read at 450 nm. This kit offers ultra-sensitive measurement of as little as 50pg/ml of IGF-1.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 10 concentrate, Standard, Wash buffer concentrate, TMB substrate, Stop solution 2, Neutralizing reagent 2, 2N HCl.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty