Would you like to log in again?
[pSer78]HSP27 (human), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-165
89141-244EA 561.14 USD InStock
89141-244
[pSer78]HSP27 (human), ELISA Kit, Enzo Life Sciences
Assays ELISAs
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.
For the quantitative determination of human [pSer78]HSP27 in cell lysates, plasma, and serum.
The [pSer78]HSP27 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 27, Wash buffer concentrate, Standard, TMB Substrate, 5X Extraction reagent, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty