Would you like to log in again?
IL-33 (human), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-201
10049-222EA 686.43 USD InStock
10049-222
IL-33 (human), ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of human IL-33 in cell lysates, culture supernatants, serum, plasma and synovial fluid samples.
The IL-33 (human), EIA kit is a colorimetric immunometric immunoassay kit with results in 3 hours.
Ordering information: Contains Microtiter Plate, Conjugate, Antibody, Assay Buffer 13, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Extraction Reagent Concentrate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty