Would you like to log in again?
Vacuum Manifold, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-987-500
10049-786EA 558.45 USD InStock
10049-786
Vacuum Manifold, Enzo Life Sciences
Manifolds
Optimized for the vacuum filtration of multi-well plates
The manifold is constructed of a chemical-resistant polymer and is designed to facilitate the filtration of liquid through a multi-well filter plate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty