Would you like to log in again?
Tegaderm™ Transparent Film Dressing, Frame Style, 3M™
Supplier: 3M Healthcare
Tegaderm™
MMM9506W MMM1624W
10017-192CS 445.1 USD InStock
10017-192 56222-191
Tegaderm™ Transparent Film Dressing, Frame Style, 3M™
First Aid Supplies Wound Dressings
Tegaderm™ Transparent film dressing film provides a barrier to external contaminants, with comprehensive clinical documentation.
A breathable dressing, impermeable to liquids, bacteria, and viruses. Picture-frame delivery style is preferred by caregivers. Secure adhesion and gentle to the skin, this breathable film allows oxygen in and moisture vapor out. Laboratory testing has proven 3M™ Tegaderm™ and Tegaderm™ HP Transparent Dressings provide a viral (HIV-1 and HBV) barrier, while dressings remain intact without leakage.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty