Would you like to log in again?
Comply™ Thermalog Steam Chemical Integrator, 3M™
Supplier: 3M
MMM2134MM
34010-086CS 653.2 USD InStock
34010-086
Comply™ Thermalog Steam Chemical Integrator, 3M™
Sterilization Indicators
Class 5 steam chemical integrators can be used in all 250 to 275 °F steam sterilization cycles.
  • Immediate and accurate method of monitoring the sterilization
  • Ink technology provides easy to read safe or unsafe result
Certifications: ANSI/AAMI/ISO 11140-1:2005 Class 5.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty