Would you like to log in again?
Hydroponics Center
Supplier: DIVERSIFIED WOODCRAFTS INC SE
HGC-52
470161-396EA 2782.3 USD InStock
470161-396
Hydroponics Center
Testing Chambers Plant Growth Chambers
Perfect For Large Classes And Greenhouses.
  • All Components Included
  • Moveable
  • Self Contained On A Single Cart
Self contained ebb and flow hydroponics growing system. The perfect way to learn how plants can grow without soil and helps teach the basic concept for the plants needs, nutrition, food production, recycling, agricultural technology and soil-less growing environments. This mobile hydroponics center includes grow lights and water pump. The adjustable timers enable you to grow your plants with the perfect amount of water and sunlight. The construction allows easy access for students and teachers alike. The containment plant tray is 8"Hx27-1/2"Dx42"W. There is a 25 gallon nutrient reservoir with lid, a locking cabinet for timers and air pump, and a 150 watt grow light. The growing system is complete with a submersible pump, an air pump, an air stone, (2) timers, hoses and 8" net pots. The unit moves easily on 4" locking, leveling casters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty