Would you like to log in again?
Ceramic Tubes, AL 23, for Heating Coils, Thermo Scientific
Supplier: Thermo Scientific
33205-QAT 33209-DM 33210-DW 33210-DR 33209-DM 33210-DW 33205-DW 33210-ED 33210-DM 33205-DR 33209-DW 33209-ED 33209-QAT 33210-QAT 33209-DW 33209-QAT 033205-DR 33205-QAT 33205-ED 33205-ED 33210-QAT 33209-DR 33210-ED 33210-DR 33209-DR 33205-DM 033205-DM 33205-DW 33209-ED 33210-DM
AA33210-EDEA 2461.98 USD InStock
AA33210-ED CAAA33210-DR CAAA33210-QAT CAAA33210-DW CAAA33210-DM AA33209-DM AA33210-QAT CAAA33205-ED AA33210-DR AA33205-DM CAAA33210-ED AA33210-DW CAAA33209-DM CAAA33205-DW CAAA33205-DR CAAA33209-DR AA33210-DM AA33209-ED CAAA33209-DW CAAA33205-DM AA33205-DW CAAA33209-QAT CAAA33205-QAT AA33205-DR CAAA33209-ED AA33209-DR AA33209-DW AA33205-ED AA33205-QAT AA33209-QAT
Ceramic Tubes, AL 23, for Heating Coils, Thermo Scientific
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL23.
  • Corrosion resistant
  • Permeable to microwaves
Dense, excellent thermal and electrical resistance properties.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty