Would you like to log in again?
VWR® Acrylic Dispenser for Parafilm Sealing Film
Supplier: Avantor
VWR®
HS234524V
82024-547EA 182.49 USD InStock
82024-547
VWR® Acrylic Dispenser for Parafilm Sealing Film
Sealing Film Dispensers
Strong, crystal-clear acrylic dispenser makes film supply visible and features a serrated cutting blade.
  • Heavy-duty construction for increased durability
  • Dispenser holds one or two 50 mm (2") rolls, or one 100 mm (4") roll
Store, dispense, and cut Parafilm, tape, labels, and other compatible rolls.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty