Would you like to log in again?
GPI Anatomicals® Cornea Eye Model
Supplier: GPI ANATOMICALS
2780
470160-254EA 135.3 USD InStock
470160-254
GPI Anatomicals® Cornea Eye Model
Models Human Biology Models
Oversized normal eye model with cut-away to show inner anatomy. Includes four interchangeable corneas that show various cornea conditions including: bullous keratopathy, Fuch’s endothelial dystrophy, keratoconus and normal. Lens and cornea are removable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty