Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Riboflavine-5'-phosphate sodium salt dihydrate ≥73.0%, Powder

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogenphosphate dihydrate

J66949-18 J66949-09
AAJ66949-09EA 47.68 USD InStock
AAJ66949-09 AAJ66949-18
Riboflavine-5'-phosphate sodium salt dihydrate ≥73.0%, Powder
Riboflavine-5'-phosphate sodium salt dihydrate
Riboflavin-5'-phosphate sodium salt dihydrate


Formula: C₁₇H₂₀N₄NaO₉P·2H₂O
MW: 514.36 g/mol (478.34 anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150993
CAS Number: 6184-17-4
EINECS: 204-988-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty