Would you like to log in again?
Nitrile Gloves, Unlined, Industrial Grade, MCR Safety
Supplier: MCR Safety
5300S 5300L 5300M 5300XL
80081-442PK 60.64 USD InStock
80081-442 80081-440 80081-436 80081-438
Nitrile Gloves, Unlined, Industrial Grade, MCR Safety
Gloves
These durable nitrile gloves contain no latex proteins and offer excellent resistance to punctures and tears.
  • Excellent alternative for individuals who experience allergic reactions to latex
Nitrile gloves are three times more puncture resistant than rubber and can be used to offer superior resistance to many types of chemicals.
Ordering information: These gloves are polybagged by the pair.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty