Would you like to log in again?
Freezer Box, Laminated Cardboard, Sarstedt
Supplier: Sarstedt
95.064.949
101093-128PK 79.19 USD InStock
101093-128
Freezer Box, Laminated Cardboard, Sarstedt
Boxes Boxes for Tubes
Freezer boxes manufactured from durable moisture-resistant laminated fiberboard
  • Tube Ø: 17 mm maximum
This box is ideal for sample storage in mechanical freezers and refrigerators. With lid and insert.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty