Would you like to log in again?
Disposable Square Cuvettes, Sarstedt
Supplier: Sarstedt
67.754 67.755
101093-996PK 21.48 USD InStock
101093-996 101093-930
Disposable Square Cuvettes, Sarstedt
Cuvettes Spectrophotometer Cuvettes
Disposable, high-quality, square plastic cuvettes are made from polystyrene (PS)
Due to the optical characteristics of the material, polystyrene cuvettes are suitable for use down to 330nm, with a spectral transmission of 70–80% at 334nm.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty