Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide, γ-phase ≥99.997% (metals basis), powder

Supplier: Thermo Scientific Chemicals

047202-14 047202-22 047202-36
AA47202-14EA 56.79 USD InStock
AA47202-14 AA47202-36 AA47202-22
Aluminium oxide, γ-phase ≥99.997% (metals basis), powder
Aluminium oxide, γ-phase
Aluminum oxide, ^y-phase, 99.997% (metals basis). 2.5 Micron Powder, S.A. 110-125m^2/g.


Formula: Al₂O₃
MW: 101.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Order Now
Specification Test Results
Assay Minimum 99.997

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty