Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Fmoc-4-(trifluoromethyl)-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-Phe(4-CF3)-OH

H66549-06 H66549-03 H66549-MD
AAH66549-03EA 150.99 USD InStock
AAH66549-03 AAH66549-MD AAH66549-06
N-Fmoc-4-(trifluoromethyl)-L-phenylalanine 95%
N-Fmoc-4-(trifluoromethyl)-L-phenylalanine


Formula: C₂₅H₂₀F₃NO₄
MW: 455.433 g/mol
Melting Pt: 127…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00797581
CAS Number: 247113-86-6
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty