Would you like to log in again?
Ferrules, Thermo Scientific
Supplier: Thermo Scientific
290GP130 290GP132 29033405 290GP131
10055-922EA 61.34 USD InStock
10055-922 10055-924 10055-920 10055-062
Ferrules, Thermo Scientific
Fittings and Connectors
Thermo Scientific ferrules are available in three different materials and various dimensions to match the instrument and capillary column I.D
Thermo Scientific 100% graphite ferrules are made of a soft material that is porous to oxygen, making them suitable for most applications except GC/MS interface connections. These easy-to-use ferrules form a soft grip with the column and provide a stable seal.
The choice of material is dependent upon the use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty