Would you like to log in again?
HyperSep™ UniGuard Direct-Connect Online SPE Columns, Thermo Scientific
Supplier: Thermo Scientific
HyperSep
60311-402 60311-301 60311-401 60311-202 60311-201 60311-502 60311-501 60311-302
10048-548EA 510.53 USD InStock
10048-548 10048-558 10048-560 10048-552 10048-550 10048-546 10048-556 10048-554
HyperSep™ UniGuard Direct-Connect Online SPE Columns, Thermo Scientific
Chromatography Columns
Clean-up and concentrate target analytes in minutes with Thermo Scientific HyperSep Online SPE products.
HyperSep UniGuard Direct-Connect Online SPE Columns attach directly to the switching valve, just like an HPLC column. They are available with the following HyperSep SPE chemistries: HyperSep Retain PEP, for retention of polar and nonpolar analytes; HyperSep Retain-CX, for retention of basic and nonpolar analytes; HyperSep Retain-AX, for retention of acidic and nonpolar analytes; and HyperSep Hypercarb, for retention of extremely polar analytes.
Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems, columns and appropriate switching valves, they provide effective removal of contaminants (such as proteins) from samples, and fast, effective clean-up and concentration of target compounds. Thermo Scientific HyperSep UniGuard Direct-Connect Online SPE Columns offer quick, easy connection by means of a reusable drop-in guard cartridge holder.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty