Would you like to log in again?
Innovative Style Goggle, Clear Anti-Fog Lens, Elastic Strap, MCR Safety
Supplier: MCR Safety
PGX122AF PGX120AF PGX112AF
89092-666EA 178.85 USD InStock
89092-666 89092-671 76264-514
Innovative Style Goggle, Clear Anti-Fog Lens, Elastic Strap, MCR Safety
Goggles
Consumers committed to sourcing premium products take confidence with PGX1®.
Four (4) styles feature dielectric design elements that include polycarbonate lens and body with thermal plastic rubber (TPR) seal,multiple points of indirect venting,external liquid diversion channel, and internal perspiration channel. Two adjustable strap options are available including elastic or TPR as well as two lens offerings in clear or gray. Additional features include anti-fog coating and microfiber cleaning / storage bag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty