Would you like to log in again?
Tribal® V Graphite Frame, MCR Safety
Supplier: MCR Safety
TB110AF TB11G TB148B TB120AF
89074-212EA 38.76 USD InStock
89074-212 89074-188 89074-180 89074-186
Tribal® V Graphite Frame, MCR Safety
Safety Glasses
The Tribal® and Tribal® V offer distinctive eyewear protection.
Multiple frame colors and a wide offering of lens color options are available for both styles. This modern and stylish design makes this a very attractive option that will be worn and keep workers safe. The Tribal® and Tribal® V meets or exceeds ANSI Z87+ and meets military MIL-PRF-31013 clause 3.5.1.1 for high velocity impact protection.
Ordering information: Cord is included with purchase.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty