Would you like to log in again?
AzureSpot Analysis Software, Azure Biosystems
Supplier: Azure Biosystems
AC1061
10147-224EA 2302.47 USD InStock
10147-224
AzureSpot Analysis Software, Azure Biosystems
Software Laboratory Software
AzureSpot helps you interpret your data easily and accurately.
  • Generate a PDF report of experiment when are done
  • Exports data in many formats to other applications (e.g. Excel) for further analysis
Software not avalible for sale without purchasing Azure Biosystems cSeries instrument.
Analyze your gels and blots in either manual or automatic modes. AzureSpot can automatically detect lanes and bands, even on distorted gels, and apply your selected method of background correction.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty