Would you like to log in again?
HyPURITY™ Aquastar 3 and 5 µm Columns, Thermo Scientific
Supplier: Thermo Scientific
HyPURITY Aquastar
22503-104030 22503-013001 22505-154630 22503-153030 22505-254630 22505-052130 22503-154630 22503-012101
10045-168EA 1248.69 USD InStock
10045-168 10045-158 10045-202 10045-184 10045-174 10045-198 10045-176 10045-156
HyPURITY™ Aquastar 3 and 5 µm Columns, Thermo Scientific
Chromatography Columns
A new member of the HyPURITY family, HyPURITY Aquastar columns are a new silica-based polar endcapped C18 designed to offer superior retention of polar compounds and increased sensitivity in 100% aqueous conditions.
HyPURITY Aquastar columns combine a C18 ligand with polar endcapping to produce a highly stable material for Reversed phase chromatography liquid chromatography. HyPURITY Aquastar columns have the unique ability to retain and separate polar analytes in 100% aqueous conditions with no phase collapse. Selectivity and sensitivity can be maintained at decreased levels of buffer concentrations, making HyPURITY Aquastar ideal for applications involving MS detection.
The robust nature of the bonded phase results in a silica-based column that is unsurpassed for LC and LC/MS applications. This next generation column also offers unmatched batch reproducibility over first generation polar endcapped columns.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty