Would you like to log in again?
AquaSil™ C18 3 and 5 µm HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
AquaSil™
77505-152130 77505-154630 77505-013001 77505-054630 77503-014001 77505-254030 77503-153030 77503-013001 77503-103030 77505-153030 77503-154630 77503-152130 77503-052130 77505-154030 77505-034030 77503-154030 77505-014001 77505-102130 77505-052130 77503-054630 77503-104630 77505-012101 77505-253030 77505-254630 77503-102130 77505-124030 77505-104630 77503-012101
10043-902EA 792.25 USD InStock
10043-902 10043-924 10043-920 10046-584 10046-580 10046-600 10046-546 10046-544 10046-588 10043-934 10043-936 10043-914 10046-608 10046-606 10043-898 10046-604 10046-594 10046-572 10046-550 10046-590 10046-556 10046-554 10043-904 10043-926 10046-598 10046-576 10046-596 10043-928
AquaSil™ C18 3 and 5 µm HPLC Columns, Thermo Scientific
Chromatography Columns
These Thermo Scientific polar endcapped C18 phase columns offer alternative selectively compared to conventional C18 phases.
Retention of polar compounds. Compatible with 100% aqueous mobile phases.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty