Would you like to log in again?
Fluophase Columns, Thermo Scientific
Supplier: Thermo Scientific
Fluophase®
82505-154630 82705-254630 82705-153030
10043-946EA 1224.04 USD InStock
10043-946 10043-974 10043-982
Fluophase Columns, Thermo Scientific
Chromatography Columns
The perfluorinated phases provide unique selectivity.
Extra retention and selectivity for halogenenated compounds. Ideal for taxane analysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty