Would you like to log in again?
BetaMax Columns, Thermo Scientific
Supplier: Thermo Scientific
Betamax
95005-254630 95205-012101 95205-052130 95205-102130
10043-996EA 657.22 USD InStock
10043-996 10043-998 10044-000 10043-990
BetaMax Columns, Thermo Scientific
Chromatography Columns
Featuring an ultra-high surface coverage for maximum retention and three Thermo Scientific chemistries designed for acid, base and neutral analytes.
  • Excellent stability for long column lifetimes
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty