Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CEA Mouse Monoclonal Antibody [clone: CEA-9]

Supplier: Boster
MA1023
10206-194EA 809.04 USD InStock
10206-194
Anti-CEA Mouse Monoclonal Antibody [clone: CEA-9]
Antibodies
Mouse IgG monoclonal antibody for CEA, carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1) detection. Tested with WB, IHC-P in Human. No cross reactivity with other proteins.

Carcinoembryonic antigen is a complex immunoreactive glycoprotein with a molecular weight of 180,000 comprising 60% carbohydrate. It is found in adenocarcinomas of endodermally derived digestive system epithelia and in fetal colon.Carcinoembryonic antigen is one of the most widely used tumor markers in serum immunoassay determinations of carcinoma.

Mouse ascites fluid, 1.2% sodium acetate, 2 mg BSA, with 0.01 mg NaN₃ as preservative.

Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: CEA
Clonality: Monoclonal
Clone: CEA-9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty