Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IKK alpha Rabbit Polyclonal Antibody

Supplier: Boster
PA1614
10209-426EA 809.04 USD InStock
10209-426
Anti-IKK alpha Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Inhibitor of nuclear factor kappa-B kinase subunit alpha(CHUK) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: IKK alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty