Would you like to log in again?
BetaSil™ Phenyl/Hexyl 3 and 5 µm HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
BetaSil™
73005-152130 73003-153030 73003-052130 73003-103030 73005-012101 73003-154630 73005-254630 73003-053030 73005-154630
10046-518EA 1376.94 USD InStock
10046-518 10046-538 10046-536 10046-512 10046-524 10046-502 10046-500 10046-520 10046-542
BetaSil™ Phenyl/Hexyl 3 and 5 µm HPLC Columns, Thermo Scientific
Chromatography Columns
BetaSil Phenyl/Hexyl columns offer a wide range of Thermo Scientific chemistries on 100 Å silica.
  • Provides unique selectivity
Featuring a high surface area with high bonded phase coverage.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty