Would you like to log in again?
HyPURITY™ C8 5 µm HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
HyPURITY™
22205-254630 22205-054630 22205-014001 22205-012101 22205-154630 22205-104630
10040-992EA 533.29 USD InStock
10040-992 10045-114 10045-128 10045-118 10045-108 10045-134
HyPURITY™ C8 5 µm HPLC Columns, Thermo Scientific
Chromatography Columns
Thermo Electron Corporation is dedicated to the design and manufacture of high quality HPLC media and columns.
The quality assurance program has a variety of highly diagnostic chromatographic tests for the investigation of both primary and secondary interactions, ensuring exceptional column-to-column and batch-to-batch reproducibility. The range of stationary phases bonded to the HyPURITY silica includes C18, C8, C4, Cyano and the unique HyPURITY ADVANCE and HyPURITY AQUASTAR phases.
Over 20 years of experience has resulted in the development of an excellent quality program and certification with ISO9001:2000. The HyPURITY family, launched in 1995, has established new standards in column characterization and validation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty