Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCMA Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-306-100
10050-274EA 547.49 USD InStock
10050-274
Anti-BCMA Rabbit Polyclonal Antibody
Antibodies
Members of the TNF superfamily regulate immune responses and induce apoptosis. Two members of this family were identified and designated BAFF and April, respectively. Three receptors for BAFF were identified; these include: BCMA (B-cell maturation protein), TACI, and BAFF-R. BCMA, like TACI, binds BAFF and April. BAFF and its receptors induce B-Cell development and survival, activate NF-κB and c-jun N-terminal kinase, and are involved in B-Cell associated autoimmune diseases.

Type: Primary
Antigen: BCMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-Terminus
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty