Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LTC4 Mouse Monoclonal Antibody [clone: 6E7]

Supplier: Enzo Life Sciences
905-902-0200
10051-942EA 593.9 USD InStock
10051-942
Anti-LTC4 Mouse Monoclonal Antibody [clone: 6E7]
Antibodies
Leukotrienes are metabolic products of arachidonic acid formed by the action of 5-lipoxygenase (5-LO), with the principal products being leukotrienes C4 (LTC4) and B4 (LTB4). Leukotriene B4 (LTB4) stimulates leukocyte functions including lysosomal enzyme release, adhesion, and aggregation of polymorphonuclear leukocytes. LTB4 has been implicated as a potent mediator of inflammatory diseases and immunoregulation. The cysteinyl leukotrienes (LTC4, LTD4, and LTE4) are formed by addition of cysteine derivatives to LTA4. The conversion of LTA4 to LTC4 by the enzyme LTC4 synthase limits the rate of cysteinyl leukotriene formation. Cysteinyl leukotrienes are potent lipid mediators in inflammation, inducing smooth muscle contractions and increased capillary permeability.

Type: Primary
Antigen: LTC4
Clonality: Monoclonal
Clone: 6E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty