Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BID Mouse Monoclonal Antibody [clone: 5C9]

Supplier: Enzo Life Sciences
ADI-AAM-141-E
10051-968EA 388.28 USD InStock
10051-968
Anti-BID Mouse Monoclonal Antibody [clone: 5C9]
Antibodies
BID is an approximately 22 kDa protein belonging to the Bcl-2 family. BID is a cytosolic protein that is cleaved by caspase-8 in response to death receptor stimulation. Proteolytic cleavage of BID results in the formation of a 15 kDa fragment, referred to as tBID. tBID migrates to the mitochondria where it aids in the release of cytochrome c and the subsequent activation of the apoptosome. tBID serves as a link between death receptor dependent and mitochondrial mediated apoptosis.

Type: Primary
Antigen: BID
Clonality: Monoclonal
Clone: 5C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty