Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF10D Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-AAP-371-E
10049-816EA 497.69 USD InStock
10049-816
Anti-TNFRSF10D Rabbit Polyclonal Antibody
Antibodies
Decoy receptors (DcRs) are signaling-deficient receptors for certain members of the TNF family of cytokines, including TRAIL/Apo2L and FasL. Functional TRAIL receptors such as DR4 and DR5 transmit apoptotic signals via their intracellular death domains. In contrast, three identified TRAIL decoy receptors (DcR1, DcR2, and osteoprotegerin/OPG) completely or partially lack the signaling death domain, and/or are secreted from cells and unable to transmit intracellular signals. Thus, expression of DcRs promotes survival by sequestration of apoptotic ligands. A fourth identified decoy receptor, DcR3, binds both FasL and the vascular endothelial cell growth inhibitor (VEGI), inhibiting Fas and DR3 death signals.

Type: Primary
Antigen: TNFRSF10D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty