Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRAF6 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-AAP-426-E
10049-818EA 374.72 USD InStock
10049-818
Anti-TRAF6 Rabbit Polyclonal Antibody
Antibodies
TRAF proteins are multifunctional signal adaptors that are involved in the activation of the kinase cascades which activate c-Jun N-terminal kinase, p38 mitogen activated protein kinase, and the transcription factor NF-κB. Members of the TRAF family form homodimers as well as heterodimers, resulting in extensive diversity and specificity to their function in signal transduction pathways.

Type: Primary
Antigen: TRAF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty