Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HMGB1 Mouse Monoclonal Antibody [clone: KS1]

Supplier: Enzo Life Sciences
ADI-CSA-614-E
10049-856EA 410.96 USD InStock
10049-856
Anti-HMGB1 Mouse Monoclonal Antibody [clone: KS1]
Antibodies
The chromatin non-histone high mobility group of proteins (HMG)-1 and -2 contain conserved domains known as HMG-boxes. Both HMG-1 and HMG-2 contain an N-terminal HMG box, a central HMG box and an acidic carboxy terminus. HMG-1/2 have been shown to facilitate the binding of various sequence-specific transcription factors to their respective DNA binding sites, such as steroid hormone receptors, and p53.

Type: Primary
Antigen: HMGB1
Clonality: Monoclonal
Clone: KS1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty