Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TAPBP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-CSA-630-D
95042-920EA 200.69 USD InStock
95042-920
Anti-TAPBP Rabbit Polyclonal Antibody
Antibodies
Tapasin is a 48 kDa glycoprotein which is a member of the immunoglobulin superfamily and is required for efficient peptide binding to TAP (Transporter associated with antigen processing). TAP binds peptides in its cytosolic part and subsequently translocates the peptides into the lumen of the endoplasmic reticulum where assembly of MHC class I and peptide takes place.

Type: Primary
Antigen: TAPBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty